Learn More
MSC MCR Safety Gloves, Goatskin, ANSI A5, Medium, 1 Pair
Catalog No. 5000676083
Encompass
These goatskin gloves offer robust cut, puncture, and abrasion resistance for demanding work environments. They feature a textured grip and are lined with Dyneema Diamond Technology Fiber to enhance protection. Designed with a hi-visibility orange back and an open cuff with a pull tab for personalized fit.
  • Lined with Dyneema Diamond Technology Fiber for enhanced cut resistance
  • Hi-visibility orange back for increased safety
  • Open cuff with pull tab and ID panel for user personalization
  • Premium goatskin palm with textured grip
  • Wing thumb with reinforced thumb crotch
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 3
  • Medium glove size (numeric size 8)
Catalog No. 50-006-76083 Supplier MSC Supplier No. 39028220
These goatskin gloves offer robust cut, puncture, and abrasion resistance for demanding work environments. They feature a textured grip and are lined with Dyneema Diamond Technology Fiber to enhance protection. Designed with a hi-visibility orange back and an open cuff with a pull tab for personalized fit.
  • Lined with Dyneema Diamond Technology Fiber for enhanced cut resistance
  • Hi-visibility orange back for increased safety
  • Open cuff with pull tab and ID panel for user personalization
  • Premium goatskin palm with textured grip
  • Wing thumb with reinforced thumb crotch
  • ANSI/ISEA cut resistance level A5
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 3
  • Medium glove size (numeric size 8)