Learn More
MSC MCR Safety Gloves, Goatskin, Padded Palm Grip, 2X-Large, 1 Pair
Catalog No. 5000677872
Encompass
These general purpose gloves are designed for various applications, offering protection against impact, abrasion, heat, and puncture. Constructed from supple grain goatskin leather with a padded palm grip, they provide comfort and enhanced control.
  • Designed for impact, abrasion, heat, and puncture protection
  • Supple grain goatskin leather is soft and comfortable
  • Padded palm grip for enhanced comfort and control
  • Thermoplastic elastomer (TPR) back provides level 1 impact protection
  • Slip-on leather cuff
  • Full-fingered with keystone thumb type
  • Shirred elastic back
  • Maximum operating temperature: 284°F
  • ANSI/ISEA abrasion resistance level 3
Catalog No. 50-006-77872 Supplier MSC Supplier No. 13352182
These general purpose gloves are designed for various applications, offering protection against impact, abrasion, heat, and puncture. Constructed from supple grain goatskin leather with a padded palm grip, they provide comfort and enhanced control.
  • Designed for impact, abrasion, heat, and puncture protection
  • Supple grain goatskin leather is soft and comfortable
  • Padded palm grip for enhanced comfort and control
  • Thermoplastic elastomer (TPR) back provides level 1 impact protection
  • Slip-on leather cuff
  • Full-fingered with keystone thumb type
  • Shirred elastic back
  • Maximum operating temperature: 284°F
  • ANSI/ISEA abrasion resistance level 3