Learn More
MSC MCR Safety Gloves, Goatskin, Padded Palm, Medium, 1 Pair
Catalog No. 5000789134
Encompass
These general purpose gloves are crafted from goatskin leather, featuring a padded palm grip and a shirred elastic back for comfort. They include a TPR back for enhanced impact protection, making them suitable for various general purpose applications.
  • Goatskin leather primary material and lining
  • Padded palm grip for enhanced handling
  • Thermoplastic rubber back for impact protection
  • Shirred elastic back for a comfortable fit
  • Slip-on cuff style
  • Full-fingered design
  • Keystone thumb type
  • ANSI/ISEA abrasion resistance level 3
  • Protection against impact, abrasion, heat, and puncture
  • Unisex design
  • Maximum operating temperature 284 °F
Catalog No. 50-007-89134 Supplier MSC Supplier No. 19940550
These general purpose gloves are crafted from goatskin leather, featuring a padded palm grip and a shirred elastic back for comfort. They include a TPR back for enhanced impact protection, making them suitable for various general purpose applications.
  • Goatskin leather primary material and lining
  • Padded palm grip for enhanced handling
  • Thermoplastic rubber back for impact protection
  • Shirred elastic back for a comfortable fit
  • Slip-on cuff style
  • Full-fingered design
  • Keystone thumb type
  • ANSI/ISEA abrasion resistance level 3
  • Protection against impact, abrasion, heat, and puncture
  • Unisex design
  • Maximum operating temperature 284 °F