Learn More
MSC MCR Safety Goatskin Gloves, Cut A5, 2X-Large, 1 Pair
Catalog No. 5000792583
Encompass
These cut, puncture, and abrasion-resistant gloves are constructed from premium goatskin with a textured grip and are beige in color. They are sold as a pair, offering robust protection for various industrial applications.
  • Lined with Dyneema Diamond Technology fiber
  • Excellent cut resistance (ANSI/ISEA A5)
  • High-visibility orange back
  • Open cuff with pull tab and ID panel for user personalization
  • Premium goatskin palm
  • Wing thumb with reinforced thumb crotch
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 3
  • Full fingered design
  • Textured grip surface
  • Slip-on cuff style
Catalog No. 50-007-92583 Supplier MSC Supplier No. 39028352
These cut, puncture, and abrasion-resistant gloves are constructed from premium goatskin with a textured grip and are beige in color. They are sold as a pair, offering robust protection for various industrial applications.
  • Lined with Dyneema Diamond Technology fiber
  • Excellent cut resistance (ANSI/ISEA A5)
  • High-visibility orange back
  • Open cuff with pull tab and ID panel for user personalization
  • Premium goatskin palm
  • Wing thumb with reinforced thumb crotch
  • ANSI/ISEA abrasion resistance level 4
  • ANSI/ISEA puncture resistance level 3
  • Full fingered design
  • Textured grip surface
  • Slip-on cuff style