Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Select Grain Unlined Cowhide Leather Drivers Gloves

Catalog No. 19058775 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-058775 Small
19-058773 Large
19-050-251 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-058775 Supplier MCR Safety Supplier No. 3211S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed from unlined select grain cowhide leather with a shirred elastic back, cotton hem, and keystone thumb pattern.

  • Select Grade Grain Cowhide
  • Keystone Thumb
  • Shirred Elastic Back
  • Color-coded hem to identify size

Specifications

ANSI Abrasion Rating ANSI 3
Back Material Shirred Elastic
Color Beige
Grade Select
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size Small
Style Gunn
For Use With (Application) Carpentry, Farming, Landscaping, Transportation, Agriculture, and Construction
Glove Cuff Style Open
Hem Color Brown
Lining Unlined
Packaging Type Polybag
Palm Type Leather
Puncture Rating ANSI/ISEA 105-16 Level 3
Series 3211
Thumb Style Keystone
Length (English) 8.6 in.
Length (Metric) 21.8 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.