Learn More
MSC MCR Safety Grip Sharp 9686, Cut A3, Medium, 1 Pair
Catalog No. 5000792443
Encompass
These Grip Sharp 9686 gloves are designed for cut, puncture, and abrasion resistance, ideal for demanding industrial applications. They feature a Kevlar primary material with a split, leather-coated palm for a textured grip. These medium-sized, 7-gauge knit gloves are yellow with a gray palm coating.
  • Knit style with seamless Kevlar lining
  • Leather-coated palm with textured grip
  • ANSI/ISEA cut resistance level A3
  • ANSI/ISEA puncture resistance level 4
  • ANSI/ISEA abrasion resistance level 5
  • Seven-gauge knit construction
  • EN 388 rating 4344
  • Reinforced thumb crotch for extended wear
  • Will not melt, ignite, or conduct electricity
  • Unisex design
Catalog No. 50-007-92443 Supplier MSC Supplier No. 79775664
These Grip Sharp 9686 gloves are designed for cut, puncture, and abrasion resistance, ideal for demanding industrial applications. They feature a Kevlar primary material with a split, leather-coated palm for a textured grip. These medium-sized, 7-gauge knit gloves are yellow with a gray palm coating.
  • Knit style with seamless Kevlar lining
  • Leather-coated palm with textured grip
  • ANSI/ISEA cut resistance level A3
  • ANSI/ISEA puncture resistance level 4
  • ANSI/ISEA abrasion resistance level 5
  • Seven-gauge knit construction
  • EN 388 rating 4344
  • Reinforced thumb crotch for extended wear
  • Will not melt, ignite, or conduct electricity
  • Unisex design