Learn More
MSC MCR Safety Hi-Viz Vest, FR Class 3, X2, 1 ea
Catalog No. 5000792844
Encompass
This flame-resistant safety vest is constructed from a mesh modacrylic/aramid blend fabric, providing Class 3 high visibility and an arc rating of 4.6 cal/cm². Designed for demanding environments, it features multiple pockets and a D-Ring access slot.
  • Two inch silver FR reflective stripes
  • Three pockets
  • ANSI/ISEA 107 Type R, Class 3 compliant
  • Arc rating 4.6 cal/cm²
  • Conforms to NFPA 70E, HRC1
  • D-Ring access for fall protection harness
  • FR hook and loop front closure
  • Hi-Vis material
  • Meets ASTM F1506
  • Mesh modacrylic/aramid blend fabric
Catalog No. 50-007-92844 Supplier MSC Supplier No. 94632783
This flame-resistant safety vest is constructed from a mesh modacrylic/aramid blend fabric, providing Class 3 high visibility and an arc rating of 4.6 cal/cm². Designed for demanding environments, it features multiple pockets and a D-Ring access slot.
  • Two inch silver FR reflective stripes
  • Three pockets
  • ANSI/ISEA 107 Type R, Class 3 compliant
  • Arc rating 4.6 cal/cm²
  • Conforms to NFPA 70E, HRC1
  • D-Ring access for fall protection harness
  • FR hook and loop front closure
  • Hi-Vis material
  • Meets ASTM F1506
  • Mesh modacrylic/aramid blend fabric