Learn More
MSC MCR Safety High Visibility Vest, Large, Breakaway, Lime, 1 ea
Catalog No. 5000767631
Encompass
This high visibility vest is designed for safety in environments requiring enhanced visibility. It features a tear-away design and is suitable for various industrial and outdoor applications.
  • Large size for comfortable fit
  • Breakaway design with hook and loop closure
  • Lime color for high visibility
  • One silver horizontal stripe, 2 in wide
  • ANSI Class 2 approved (ANSI 107-2015)
  • Constructed from mesh material
Catalog No. 50-007-67631 Supplier MSC Supplier No. 43829894
This high visibility vest is designed for safety in environments requiring enhanced visibility. It features a tear-away design and is suitable for various industrial and outdoor applications.
  • Large size for comfortable fit
  • Breakaway design with hook and loop closure
  • Lime color for high visibility
  • One silver horizontal stripe, 2 in wide
  • ANSI Class 2 approved (ANSI 107-2015)
  • Constructed from mesh material