Learn More
MSC MCR Safety Hulk HK3 Safety Glasses, Gray Anti-Fog, Frameless, 12/Pack
Catalog No. 5000788673
Encompass
The MCR Safety HK312PF Hulk HK3 Series Safety Glasses feature a foam-lined, frameless design, offering comprehensive eye protection. Equipped with soft, secure TPR nose pieces and non-slip temples, these glasses provide extended comfort and stability during wear.
  • Foam lined frameless design
  • Soft secure TPR nose piece
  • Non-slip TPR temples
  • Excellent orbital seal for eye protection
  • Closed cell foam lining, latex-free
  • Premium MAX6 anti-fog lens coating
  • Passes ANSI/ISEA Z87.1 2020 anti-fog test
  • Meets ANSI/ISEA Z87.1-2025 and CAN/CSA Z94.3 standards
  • Provides 100% UV protection
Catalog No. 50-007-88673 Supplier MSC Supplier No. 39874896
The MCR Safety HK312PF Hulk HK3 Series Safety Glasses feature a foam-lined, frameless design, offering comprehensive eye protection. Equipped with soft, secure TPR nose pieces and non-slip temples, these glasses provide extended comfort and stability during wear.
  • Foam lined frameless design
  • Soft secure TPR nose piece
  • Non-slip TPR temples
  • Excellent orbital seal for eye protection
  • Closed cell foam lining, latex-free
  • Premium MAX6 anti-fog lens coating
  • Passes ANSI/ISEA Z87.1 2020 anti-fog test
  • Meets ANSI/ISEA Z87.1-2025 and CAN/CSA Z94.3 standards
  • Provides 100% UV protection