Learn More
MSC MCR Safety Insulated Work Gloves, Latex Coated, X-Large, 1 Pair
Catalog No. 5000768115
Encompass
The MCR Safety NXG® Insulated Work Gloves feature a 10-gauge hi-visibility acrylic shell and blue latex-coated palm and fingertips. These gloves provide premium quality, comfort, and fit for demanding workdays in multi-industrial applications requiring high dexterity.
  • Fluorescent lime 10-gauge acrylic shell
  • Blue latex dip on the palm and fingertips
  • Excellent comfort and fit
  • Suitable for heavier duty requirements
  • High comfort level
  • Insulated gloves provide extra thermal protection
  • High visibility
Catalog No. 50-007-68115 Supplier MSC Supplier No. 02485332
The MCR Safety NXG® Insulated Work Gloves feature a 10-gauge hi-visibility acrylic shell and blue latex-coated palm and fingertips. These gloves provide premium quality, comfort, and fit for demanding workdays in multi-industrial applications requiring high dexterity.
  • Fluorescent lime 10-gauge acrylic shell
  • Blue latex dip on the palm and fingertips
  • Excellent comfort and fit
  • Suitable for heavier duty requirements
  • High comfort level
  • Insulated gloves provide extra thermal protection
  • High visibility