Learn More
MSC MCR Safety Insulated Work Gloves, X-Large, Hi-Vis, PVC-Coated, 1 ea
Catalog No. 5000686296
Encompass
The MCR Safety Ninja® Ice N9690HVXL gloves are X-Large, high-visibility, insulated work gloves designed for general purpose use. They feature a lime nylon shell with an acrylic terry interior liner and are over-the-knuckle coated with HPT for enhanced grip in wet or dry conditions, even in low temperatures.
  • X-Large size for a comfortable fit
  • High-visibility lime nylon shell with acrylic terry interior liner
  • Over-the-knuckle HPT coating repels liquids for wet or dry grip
  • Coating remains soft in temperatures as low as -4°F
  • Insulated acrylic terry liner keeps hands warm
  • Provides protection against abrasion, heat, cut, and puncture
  • Treated with ActiFresh® to help control bacteria and odor
  • Encapsulated air molecules provide a cushioning effect to minimize impact
Catalog No. 50-006-86296 Supplier MSC Supplier No. 43783430
The MCR Safety Ninja® Ice N9690HVXL gloves are X-Large, high-visibility, insulated work gloves designed for general purpose use. They feature a lime nylon shell with an acrylic terry interior liner and are over-the-knuckle coated with HPT for enhanced grip in wet or dry conditions, even in low temperatures.
  • X-Large size for a comfortable fit
  • High-visibility lime nylon shell with acrylic terry interior liner
  • Over-the-knuckle HPT coating repels liquids for wet or dry grip
  • Coating remains soft in temperatures as low as -4°F
  • Insulated acrylic terry liner keeps hands warm
  • Provides protection against abrasion, heat, cut, and puncture
  • Treated with ActiFresh® to help control bacteria and odor
  • Encapsulated air molecules provide a cushioning effect to minimize impact