Learn More
MCR Safety K3H Magnifier Safety Glasses
Catalog No. 19810408 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Magnification Power:
+1.50
+2.50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-810-408 +2.50
19-810-407 +1.50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-810-408 Supplier MCR Safety Supplier No. K3H25G
This item is not returnable. View return policy
The Klondike™Plus Magnifier from MCR Safety features modern style combined with excellent fit, comfort, and adjustability. The Klondike Plus Magnifier also provides 99.9% UV protection, meeting ANSI Z87+ standard with a range of magnifier options.
  • 1.0, 1.5, 2.0, and 2.5 diopters available
  • Black matte finish
  • Adjustable, 5 position ratchet action spatula temple
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Lens Coating Scratch-resistant
Lens Tint Gray
Product Type Magnifier Safety Glasses, 2.5X
Frame Color Black
Magnification Power +2.50
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400