Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Reversible Unhemmed Cotton Lisle Inspection Gloves

Catalog No. 19813569 Shop All MCR Safety Products
Change view
Click to view available options
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-813-569 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-813-569 Supplier MCR Safety Supplier No. 8610C
This item is not returnable. View return policy

Lightweight, breathable inspection gloves are constructed of 100% cotton and can be worn alone or used as glove liners.

  • 100% cotton lisle construction
  • Breathable fabric helps keep hands cool and comfortable
  • Size small designed specifically for women
  • Reversible, unhemmed
  • Straight thumb

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Material Cotton
Product Type Inspection Glove
Size Small
Style Reversible
For Use With (Application) Assembly, Clinical Laboratories, Electronic Components and Inspection
Gender Women's
Glove Cuff Style Unhemmed
Series 8610C
Thumb Style Straight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.