Learn More
MSC MCR Safety Latex-Coated Gloves, Crinkle Grip, X-Large, 1 Pair
Catalog No. 5000802487
Encompass
These general purpose work gloves are designed for a variety of applications, offering protection against abrasion, heat, cut, and puncture. They feature a latex coating on the palm and fingertips for enhanced grip.
  • Cotton glove material with acrylic lining
  • Knit cuff style
  • Crinkle grip surface
  • X-Large size
  • Wing thumb type
  • Dipped latex coating on palm and fingertips
  • Provides protection against abrasion, heat, cut, and puncture
  • Maximum temperature of 284 °F
Catalog No. 50-008-02487 Supplier MSC Supplier No. 77208577
These general purpose work gloves are designed for a variety of applications, offering protection against abrasion, heat, cut, and puncture. They feature a latex coating on the palm and fingertips for enhanced grip.
  • Cotton glove material with acrylic lining
  • Knit cuff style
  • Crinkle grip surface
  • X-Large size
  • Wing thumb type
  • Dipped latex coating on palm and fingertips
  • Provides protection against abrasion, heat, cut, and puncture
  • Maximum temperature of 284 °F