Learn More
MSC MCR Safety LCS1, Anti-Fog, Anti-Static, 600 Wipes, 8 oz
Catalog No. 5000686789
Encompass
This disposable lens cleaning station is designed for the care and maintenance of safety eyewear. It includes an 8 oz cleaning solution and 600 tissues, featuring anti-fog and anti-static properties to help keep glasses clean.
  • Provides a comprehensive lens cleaning station
  • Includes an 8 oz cleaning solution in a pump bottle
  • Comes with 600 tissues (two boxes of 300)
  • Designed for plastic, polycarbonate, and glass lenses
  • Features a silicone-free formula
  • Can be easily mounted on a wall or displayed on a table
  • Suitable for cleaning glasses, goggles, face shields, and computer screens
  • Safe and effective for all types of lens coatings
  • Offers anti-fog and anti-static performance
Catalog No. 50-006-86789 Supplier MSC Supplier No. 05183371
This disposable lens cleaning station is designed for the care and maintenance of safety eyewear. It includes an 8 oz cleaning solution and 600 tissues, featuring anti-fog and anti-static properties to help keep glasses clean.
  • Provides a comprehensive lens cleaning station
  • Includes an 8 oz cleaning solution in a pump bottle
  • Comes with 600 tissues (two boxes of 300)
  • Designed for plastic, polycarbonate, and glass lenses
  • Features a silicone-free formula
  • Can be easily mounted on a wall or displayed on a table
  • Suitable for cleaning glasses, goggles, face shields, and computer screens
  • Safe and effective for all types of lens coatings
  • Offers anti-fog and anti-static performance