Learn More
MSC MCR Safety Leather Drivers Gloves, Keystone Thumb, E Grade, 12/pk
Catalog No. 5000794975
Encompass
MCR Safety driver-style gloves are crafted from cowhide leather, known for its excellent balance of abrasion resistance, dexterity, durability, and comfort. These gloves feature a split leather back and a grain leather patch palm.
  • Competitive Value (CV) E grade
  • Split leather back
  • Grain leather patch palm
  • Keystone thumb design
  • Color-coded hem to identify size
  • Shirred elastic back
Catalog No. 50-007-94975 Supplier MSC Supplier No. 52387792
MCR Safety driver-style gloves are crafted from cowhide leather, known for its excellent balance of abrasion resistance, dexterity, durability, and comfort. These gloves feature a split leather back and a grain leather patch palm.
  • Competitive Value (CV) E grade
  • Split leather back
  • Grain leather patch palm
  • Keystone thumb design
  • Color-coded hem to identify size
  • Shirred elastic back