Learn More
MCR Safety Lens Cleaning Station
Catalog No. 19041832 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Description:
Lens Cleaning Station
Catalog No. Description
19-041832 Lens Cleaning Station
1 options
Catalog No. 19-041832 Supplier MCR Safety Supplier No. LCS1
Add to Cart
Add to Cart
MCR Safety offers a wide variety of products to assist in the care and maintenance of your safety eyewear. From cases, cords, and everything in between we have you covered for all your eyewear accessories needs. Be accessorized with MCR Safety!
  • Cleaning Station
  • 8 Ounce Spray Cleaning Solution
  • 2 Boxes of 300 Tissues
Specifications
Description Lens Cleaning Station
Product Type Cleaning Station
For Use With (Equipment) For plastic, polycarbonate glasses and lens
For Use With (Application) Reduces fogging/static cling
Includes 2 boxes of 300 tissues; 1.8 oz. anti-fog/anti-static spray