Learn More
MSC MCR Safety Lens Towelettes, Antifog, Antistatic, Silicone Free, 100 Wipes/Box
Catalog No. 5000782008
Encompass
These lens cleaning towelettes are designed for the care and maintenance of safety eyewear. They are safe and effective for cleaning various lens coatings, helping to keep safety glasses clean and workers injury-free.
  • Each box contains 100 individually wrapped wipes
  • For plastic, polycarbonate, and glass lenses
  • Antifog, antistatic, pre-moistened
  • Silicone free
Catalog No. 50-007-82008 Supplier MSC Supplier No. 71900708
These lens cleaning towelettes are designed for the care and maintenance of safety eyewear. They are safe and effective for cleaning various lens coatings, helping to keep safety glasses clean and workers injury-free.
  • Each box contains 100 individually wrapped wipes
  • For plastic, polycarbonate, and glass lenses
  • Antifog, antistatic, pre-moistened
  • Silicone free