Learn More
MSC MCR Safety Magnifying Safety Glasses, 2.00 Magnification, Gray, 1 ea
Catalog No. 5000795986
Encompass
Magnifying Safety Glasses provide visual enhancement with 2.00 magnification. They feature gray, scratch-resistant lenses and meet ANSI Z87.1+ safety standards. The glasses are designed with black, full-framed frames and offer UV protection.
  • Magnification of 2.00
  • Gray lenses with scratch-resistant properties
  • Meets ANSI Z87.1+ standards for eye protection
  • Black, full-framed design
  • Provides UV protection
Catalog No. 50-007-95986 Supplier MSC Supplier No. 08517120
Magnifying Safety Glasses provide visual enhancement with 2.00 magnification. They feature gray, scratch-resistant lenses and meet ANSI Z87.1+ safety standards. The glasses are designed with black, full-framed frames and offer UV protection.
  • Magnification of 2.00
  • Gray lenses with scratch-resistant properties
  • Meets ANSI Z87.1+ standards for eye protection
  • Black, full-framed design
  • Provides UV protection