Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Bobcat™ Jersey-Lined Premium Select Side-Split Leather Welding Gloves

Catalog No. 19058699 Shop All MCR Safety Products
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-058-699 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-058-699 Supplier MCR Safety Supplier No. 4740
This item is not returnable. View return policy

Gloves feature premium select side-split cow leather construction with a flame-resistant cuff liner. Sewn with Aramid Fiber for maximum protection when exposed to heat, flames, or sparks.

  • Premium select side-split cow leather
  • Sewn with Aramid Fiber
  • Rolled gauntlet cuff with flame resistant liner
  • Reinforced straight thumb
  • Fully welted

Specifications

Cuff Material Flame Resistant
Color Natural
Grade Premium
Material Side Split Cowhide Leather
Size X-Large
Style Gunn
For Use With (Application) Exposure to Heat, Flames, and Sparks
Glove Cuff Style Gauntlet
Liner Material Jersey
Series 4740
Thumb Style Straight
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.