Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Artic Jack™ Thermosock™-Lined Split Pigskin Leather Palm Work Gloves

Catalog No. 19058697 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-058-697 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-058-697 Supplier MCR Safety Supplier No. 1956XL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully lined with Thermosock™ lining to keep hands warm during cold storage, or below freezing applications. Gloves feature a split pigskin palm and fingers with a blue fabric shirred elastic back.

  • Split pigskin leather
  • Blue fabric back
  • Shirred elastic back
  • Thermosock™ lining
  • Elastic wrist

Specifications

Color Natural, Blue
Grade Premium
Hand Specific
Material Pigskin Leather
Product Type Thermal Lined Gloves
Size X-Large
Style Gunn
Glove Cuff Color Blue
Glove Cuff Style Knit
Hem Color Blue
Liner Material Thermosock™
Packaging Type Polybag
Series Artic Jack™ 1956
Thumb Style Wing
Length (English) 10.7 in.
Length (English) Cuff 2.5 in.
Length (Metric) 27.2 cm
Length (Metric) Cuff 6.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.