Learn More
MSC MCR Safety Mechanics Gloves, Goatskin, ANSI Cut A5, 1 Pair
Catalog No. 5000769065
Encompass
The UltraTech mechanics glove provides robust protection against cuts, punctures, and abrasion. It features a premium goatskin palm lined with Dyneema Diamond Technology fiber for superior cut resistance, a hi-visibility orange back, and a reinforced thumb crotch. Designed with an open cuff and ID panel for user personalization, these gloves meet ANSI Cut A5, ANSI Puncture 3, and ANSI Abrasion 4 standards.
  • Lined with Dyneema Diamond Technology fiber
  • Excellent cut resistance
  • Hi-visibility orange back
  • Open cuff with pull tab and ID panel for user personalization
  • Premium goatskin palm
  • Wing thumb with reinforced thumb crotch
Catalog No. 50-007-69065 Supplier MSC Supplier No. 39028287
The UltraTech mechanics glove provides robust protection against cuts, punctures, and abrasion. It features a premium goatskin palm lined with Dyneema Diamond Technology fiber for superior cut resistance, a hi-visibility orange back, and a reinforced thumb crotch. Designed with an open cuff and ID panel for user personalization, these gloves meet ANSI Cut A5, ANSI Puncture 3, and ANSI Abrasion 4 standards.
  • Lined with Dyneema Diamond Technology fiber
  • Excellent cut resistance
  • Hi-visibility orange back
  • Open cuff with pull tab and ID panel for user personalization
  • Premium goatskin palm
  • Wing thumb with reinforced thumb crotch