Learn More
MSC MCR Safety Mechanics Lifting Gloves, Medium, Goatskin Leather, 1 Pair
Catalog No. 5000803071
Encompass
These MCR Safety Mechanic's & Lifting Gloves are thermosock-lined, white and lime green, and designed for high visibility. They feature reflective stripes on the back and fingertips, an adjustable hook and loop closure, and are constructed from premium grain goatskin leather for durability and comfort.
  • Thermosock lining provides insulation
  • High visibility material with quality reflective stripes
  • Two reflective stripes on back and three fingertips
  • Adjustable hook and loop closure for a secure fit
  • Premium grain goatskin leather construction
  • Textured grip surface for enhanced handling
  • Provides abrasion and puncture protection
  • Keystone thumb type for improved dexterity
  • Full-fingered design
Catalog No. 50-008-03071 Supplier MSC Supplier No. 68048271
These MCR Safety Mechanic's & Lifting Gloves are thermosock-lined, white and lime green, and designed for high visibility. They feature reflective stripes on the back and fingertips, an adjustable hook and loop closure, and are constructed from premium grain goatskin leather for durability and comfort.
  • Thermosock lining provides insulation
  • High visibility material with quality reflective stripes
  • Two reflective stripes on back and three fingertips
  • Adjustable hook and loop closure for a secure fit
  • Premium grain goatskin leather construction
  • Textured grip surface for enhanced handling
  • Provides abrasion and puncture protection
  • Keystone thumb type for improved dexterity
  • Full-fingered design