Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Premium Grade Grain Cowhide Leather Drivers Gloves with Fleece Lining

Catalog No. 189992775C Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Series:
3250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Series
18-999-2775C 3250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-2775C Supplier MCR Safety Supplier No. 3250L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves feature a red fleece lining for warmth and comfort, a shirred elastic back, and a straight thumb pattern.

  • Premium Grade Grain Cowhide Leather
  • Red Fleece Lining
  • Straight Thumb
  • Shirred Elastic Back
  • Color-coded hem for size identification

Specifications

Color White
Grade Premium
Hand Specific
Material Grain Cowhide Leather
Product Type Driver Gloves
Size Large
Style Gunn
For Use With (Application) Carpentry, Cold Storage Applications, Farming, and Landscaping
Glove Cuff Style Open
Hem Color Brown
Liner Material Fleece
Packaging Type Polybag
Series 3250
Thumb Style Straight
Length (English) 10.3 in.
Length (Metric) 26.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.