Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Predator™ Nitrile Coated Work Gloves with ActiFresh™ Lining

Catalog No. 19050280 Shop All MCR Safety Products
Change view
Click to view available options
:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-050-280 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-050-280 Supplier MCR Safety Supplier No. 9761
This item is not returnable. View return policy

Nitrile-coated gloves feature a cotton jersey lining for comfort, and a safety cuff for increased protection. Acti-Fresh™ treatment helps prevent odors while also eliminating bacteria.

  • Premium nitrile coating over the entire glove
  • Soft jersey lining for increase comfort
  • Safety cuff for additional wrist protection
  • Treated with ActiFresh™ to eliminate odors and bacteria
  • Materials comply with all relevant federal regulations for food contact

Specifications

Coated Area Full Glove
Cuff Material Canvas
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Grade Premium
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Agriculture, Carpentry, Construction, Farming, Forestry, Material Handling, and Metal Fabrication
Glove Cuff Color Natural
Glove Cuff Style Safety
Packaging Type Polybag
Series Predator™
Thumb Style Straight
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.