Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Neoprene Sandpaper Grip Gloves

Catalog No. 19813566 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-566 Small
19-813-565 Medium
19-813-564 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-813-566 Supplier MCR Safety Supplier No. 6962S

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Double-dipped supported neoprene provides outstanding protection from cuts, abrasion and hazardous chemicals

  • Sandpaper finish for a secure grip
  • Maintains flexibility in cold temperatures
  • 12 in. gauntlet cuff
  • 100% cotton interlock liner
  • Launderable
  • Wing thumb

For use in chemical processing, degreasing, general maintenance, metal finishing, lubricating, and transmission assembly.

Specifications

Color Black, Blue
Material Neoprene
Size Small
For Use With (Application) chemical processing, chemical splash, degreasing, equipment cleaning, Oil and Gas industry, Oil refining
Glove Cuff Style Gauntlet
Grip Design Rough
Length (English) 12 in.
Length (Metric) 30.48 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.