Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Ninja™ X Bi-Polymer Coated Nylon Lycra™ Work Gloves

Catalog No. 19813626 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-626 Small
19-813-625 Medium
19-813-611 Large
19-813-627 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-813-626 Supplier MCR Safety Supplier No. N9674S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Bi-polymer coated gloves provide an assortment of dexterity, sense-of-touch, and protection. Actifresh™ treatment helps to prevent odors and bacteria.

  • 15 gauge Nylon Lycra™ shell increases dexterity
  • Bi-Polymer coating provides abrasion resistance
  • Actifresh™ treatments prevents odors and eliminates bacteria
  • Dark color hides dirt and grime

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Bi-Polymer
Color Black
Hand Specific
Material Nylon, Lycra
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Automotive Assembly, Landscaping, Material Handling, and Warehousing
Glove Cuff Style Knit
Series Ninja™ N9674
Yarn Gauge 15
Length (English) 12.6 in.
Length (Metric) 32 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.