Learn More
MSC MCR Safety NitriShield Gloves, 6 mil, Powder Free Nitrile, 100/Bx
Catalog No. 5000796643
Encompass
Introducing NitriShield® with Grippaz™ Technology, these single-use gloves are engineered with a patented suction grip design effective on wet and greasy surfaces. An embossed internal grip enhances traction and prevents slipping against the skin, while the lattice effect of the grip improves tensile strength and durability.
  • Powder free disposable nitrile material
  • Thickness of 6 mil (6.3 g)
  • Overall length of 9.5 in.
  • Black color
  • Textured grip with fish-scale pattern
  • Passes ASTM D6978 for fentanyl handling
  • Suitable for gastric acid fluid contact
  • Ambidextrous design
  • Touchscreen friendly
  • Latex free
  • FDA accepted material for food contact
  • Industrial and food service grade
Catalog No. 50-007-96643 Supplier MSC Supplier No. 15819444
Introducing NitriShield® with Grippaz™ Technology, these single-use gloves are engineered with a patented suction grip design effective on wet and greasy surfaces. An embossed internal grip enhances traction and prevents slipping against the skin, while the lattice effect of the grip improves tensile strength and durability.
  • Powder free disposable nitrile material
  • Thickness of 6 mil (6.3 g)
  • Overall length of 9.5 in.
  • Black color
  • Textured grip with fish-scale pattern
  • Passes ASTM D6978 for fentanyl handling
  • Suitable for gastric acid fluid contact
  • Ambidextrous design
  • Touchscreen friendly
  • Latex free
  • FDA accepted material for food contact
  • Industrial and food service grade