Learn More
MCR Safety Pants
Catalog No. 19070530 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size: X-Large
Catalog No.
19-070530 Size: X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-070530 Supplier MCR Safety Supplier No. X400PWXL
This item is not returnable. View return policy
Cylcone Series offers excellent strength and durability for tough jobs. Cyclone pants feature a wavelock pattern nylon between two layers of PVC that give it great rip resistance and abrasion resistance.
The 400PW is an elastic waist pant option with snap fly front and take up snaps on each ankle. . Limited flammability, NOT intended for use as protection against ARC Flash or Flash Fire.
  • An excellent choice for tough and rugged garments
  • .35mm PVC/Nylon/PVC, elastic waist pants
  • Snap fly front
  • Take up snaps on each ankle
  • No pockets
Specifications
Waist Style Elastic