Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Predaflex™ Fully Nitrile Coated Work Gloves

Catalog No. 19813583 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Coated Area:
Full Glove
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coated Area
19-813-583 Full Glove
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-813-583 Supplier MCR Safety Supplier No. 9792L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully coated rough nitrile finish provides durability and enhances grip capabilities, with a cotton brushed interlock liner for increased comfort.

  • Full nitrile coating features a rough finish and full flexibility
  • Brushed cotton interlock liner increases user comfort
  • ActiFresh treated to combat odor and bacteria
  • Component materials comply with all federal regulations for food contact

Specifications

Certifications/Compliance FDA
Coated Area Full Glove
Coating Material Nitrile
Color Blue
Exterior Finish Fully Textured
Grade Industrial
Hand Specific
Product Type Dip-coated Gloves
Size Large
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AKLMNT
For Use With (Application) Agriculture, Carpentry, Construction, Material Handling, Petrochemical Industries, and Oil/Gas
Glove Cuff Style Gauntlet
Grip Design Rough
Liner Material Cotton
Resistant To Abrasion, Chemicals
Series 9792
Length (English) 11.8 in.
Length (Metric) 30 cm
Product Line Predaflex
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.