Learn More
MSC MCR Safety Premium A Grade Double Leather Palm, 12 pr
Catalog No. 5000767700
Encompass
These double leather palm work gloves are constructed with two layers of premium A+ split side cowhide leather in the palm, index finger, and two middle fingers. They are sewn with super-strong, heat-resistant Kevlar® thread for demanding jobs. These gloves also feature an 8 oz. jersey palm lining, a 9 oz. cotton canvas back, and a 2.75-inch rubberized safety cuff.
  • Double leather palm construction
  • Premium A+ split side leather
  • Rubberized safety cuff, 2.75 inch
  • Double palm, index finger, and two middle fingers
  • Sewn with Kevlar® thread
  • Absorbent fleece lining, fully in-sewn on thumb and little finger
  • Cotton canvas back
Catalog No. 50-007-67700 Supplier MSC Supplier No. 29222858
These double leather palm work gloves are constructed with two layers of premium A+ split side cowhide leather in the palm, index finger, and two middle fingers. They are sewn with super-strong, heat-resistant Kevlar® thread for demanding jobs. These gloves also feature an 8 oz. jersey palm lining, a 9 oz. cotton canvas back, and a 2.75-inch rubberized safety cuff.
  • Double leather palm construction
  • Premium A+ split side leather
  • Rubberized safety cuff, 2.75 inch
  • Double palm, index finger, and two middle fingers
  • Sewn with Kevlar® thread
  • Absorbent fleece lining, fully in-sewn on thumb and little finger
  • Cotton canvas back