Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Premium Split Leather Insulated Drivers Gloves with Pile Lining

Catalog No. 19059607A Shop All MCR Safety Products
Change view
Click to view available options
:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-059-607A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-059-607A Supplier MCR Safety Supplier No. 3170S
This item is not returnable. View return policy

These split leather gloves feature an acrylic pile lining for warmth, a shirred elastic back, and straight thumb.

  • Premium Brown Split Cowhide Leather
  • Acrylic Pile Lining
  • Keystone Thumb
  • Color coded hem to identify sizes

Specifications

Color Brown
Grade Premium
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type Driver Gloves
Size Small
Style Gunn
For Use With (Application) Carpentry, Cold Storage Applications, Farming, and Landscaping
Glove Cuff Style Open
Hem Color Brown
Liner Material Acrylic Pile
Packaging Type Polybag
Series 3170
Thumb Style Keystone
Length (English) 8.8 in.
Length (Metric) 22.4 cm
Weight 41.5 lb. (Case)
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.