Learn More
Enterprise Technology Solutions MCR Safety Protective Eyewear, Single Lens, Uncoated, Clear
SDP
Catalog No. 5000827933
Encompass_Preferred
Protective eyewear provides increased angular coverage and improved peripheral vision. Molded-in browguard and vented sideshields give additional protection. Yukon Safety Glasses are made of rugged polycarbonate with no metal parts. The wraparound lens filters 99.9 percent harmful, ultraviolet rays. Uncoated eyewear meets or exceeds ANSI Z87.1 standard.
Catalog No. 50-008-27933 Supplier Enterprise Technology Solutions Supplier No. MCS9800
Protective eyewear provides increased angular coverage and improved peripheral vision. Molded-in browguard and vented sideshields give additional protection. Yukon Safety Glasses are made of rugged polycarbonate with no metal parts. The wraparound lens filters 99.9 percent harmful, ultraviolet rays. Uncoated eyewear meets or exceeds ANSI Z87.1 standard.