Learn More
MSC MCR Safety Rain Jacket, .42mm PVC/Polyester/PVC, Limited Flammability, 1 ea
Catalog No. 5000781485
Encompass
The Dominator Series rain jacket provides durable protection for demanding work environments. Constructed from .42mm PVC/hi-tensile polyester/PVC, this jacket is designed to guard against splash, grit, and grime, featuring a waterproof design and limited flammability.
  • Features .42mm PVC/hi-tensile polyester/PVC construction
  • Inner sleeves prevent splash back
  • Attached drawstring hood
  • Take-up snaps at wrists
  • Limited flammability (not for ARC Flash or Flash Fire protection)
  • Zipper front with double storm flap
  • No pockets or vents for enhanced splash protection
  • Tested in accordance with ASTM F903
Catalog No. 50-007-81485 Supplier MSC Supplier No. 43828102
The Dominator Series rain jacket provides durable protection for demanding work environments. Constructed from .42mm PVC/hi-tensile polyester/PVC, this jacket is designed to guard against splash, grit, and grime, featuring a waterproof design and limited flammability.
  • Features .42mm PVC/hi-tensile polyester/PVC construction
  • Inner sleeves prevent splash back
  • Attached drawstring hood
  • Take-up snaps at wrists
  • Limited flammability (not for ARC Flash or Flash Fire protection)
  • Zipper front with double storm flap
  • No pockets or vents for enhanced splash protection
  • Tested in accordance with ASTM F903