Learn More
MSC MCR Safety Rain Jacket, PVC/Hi-Tensile Polyester/PVC, Green, Small, 1 ea
Catalog No. 5000803741
Encompass
This rain jacket, part of the Dominator Series, is a 1-piece, waterproof garment constructed from .42mm PVC/Hi-Tensile Polyester/PVC. Designed for demanding jobs, it offers protection against splash, grit, and grime, featuring a zipper front and an attached drawstring hood.
  • Constructed from .42 mm PVC/Hi-Tensile Polyester/PVC
  • Inner sleeves prevent splash back
  • Attached drawstring hood
  • Take-up snaps at wrists
  • Limited flammability (not for ARC Flash or Flash Fire)
  • Zipper front with double storm flap
  • No pockets or vents
  • Tested in accordance with ASTM F903 for liquid penetration resistance
Catalog No. 50-008-03741 Supplier MSC Supplier No. 43828110
This rain jacket, part of the Dominator Series, is a 1-piece, waterproof garment constructed from .42mm PVC/Hi-Tensile Polyester/PVC. Designed for demanding jobs, it offers protection against splash, grit, and grime, featuring a zipper front and an attached drawstring hood.
  • Constructed from .42 mm PVC/Hi-Tensile Polyester/PVC
  • Inner sleeves prevent splash back
  • Attached drawstring hood
  • Take-up snaps at wrists
  • Limited flammability (not for ARC Flash or Flash Fire)
  • Zipper front with double storm flap
  • No pockets or vents
  • Tested in accordance with ASTM F903 for liquid penetration resistance