Learn More
MSC MCR Safety Rain Pants, High-Visibility Lime, 4X-Large, 1 ea
Catalog No. 5000795688
Encompass
MCR Safety Rain Pants are crafted from a durable blend of polyester and polyurethane, designed to offer protection in wet conditions. These pants feature a snap closure and high-visibility lime color, meeting Class E certification standards for safety.
  • Material: polyester and polyurethane blend
  • Snaps closure
  • High-visibility lime color
  • Class E certified
  • Material weight of 16.96 oz
  • No pockets
Catalog No. 50-007-95688 Supplier MSC Supplier No. 42828673
MCR Safety Rain Pants are crafted from a durable blend of polyester and polyurethane, designed to offer protection in wet conditions. These pants feature a snap closure and high-visibility lime color, meeting Class E certification standards for safety.
  • Material: polyester and polyurethane blend
  • Snaps closure
  • High-visibility lime color
  • Class E certified
  • Material weight of 16.96 oz
  • No pockets