Learn More
MSC MCR Safety Rain Suit, Hi-Visibility, 3X-Large, Lime, 1 ea
Catalog No. 5000788702
Encompass
The MCR Safety Luminator Hi-Visibility Rain Suit is a 2-piece garment engineered for superior protection in demanding outdoor environments. Constructed from a .40mm PU/cotton poly blend stretch fabric, this fluorescent lime suit offers both waterproof integrity and comfort, meeting ANSI/ISEA 107 2020 Type R Class 3 standards for high visibility.
  • Fabric: .40mm PU / cotton poly blend stretch
  • Two piece hi vis reflective rain suit
  • Meets ANSI/ISEA 107 2020 standard
  • Type R class 3 compliant
  • Stretch fabric moves for comfortable fit
  • Jacket with attached drawstring hood
  • Cape ventilated back with D-ring access slot
  • Limited flammability (tested per ASTM D-6413)
  • Zipper front with storm flap
  • Two front patch pockets
  • Elastic inner cuffs
  • Drawstring elastic waist pants
  • Expandable ankle gussets
Catalog No. 50-007-88702 Supplier MSC Supplier No. 62677984
The MCR Safety Luminator Hi-Visibility Rain Suit is a 2-piece garment engineered for superior protection in demanding outdoor environments. Constructed from a .40mm PU/cotton poly blend stretch fabric, this fluorescent lime suit offers both waterproof integrity and comfort, meeting ANSI/ISEA 107 2020 Type R Class 3 standards for high visibility.
  • Fabric: .40mm PU / cotton poly blend stretch
  • Two piece hi vis reflective rain suit
  • Meets ANSI/ISEA 107 2020 standard
  • Type R class 3 compliant
  • Stretch fabric moves for comfortable fit
  • Jacket with attached drawstring hood
  • Cape ventilated back with D-ring access slot
  • Limited flammability (tested per ASTM D-6413)
  • Zipper front with storm flap
  • Two front patch pockets
  • Elastic inner cuffs
  • Drawstring elastic waist pants
  • Expandable ankle gussets