Learn More
MSC MCR Safety Rain Suit, X-Large, ANSI Class 3, 1 ea
Catalog No. 5000788423
Encompass
This two-piece rain suit is engineered for applications requiring high visibility and protection against the elements. It features a jacket with an attached hood and elastic waist pants, designed to meet rigorous safety standards for high-visibility apparel.
  • Constructed from .40mm PU / Cotton Poly Blend stretch fabric
  • Meets ANSI/ISEA 107 2020 Type R Class 3 standards for high visibility
  • Features 2-inch silver reflective stripes
  • Jacket includes a zippered front with storm flap, two front patch pockets, and elastic inner cuffs
  • Pants feature an elastic waist with drawstring and expandable ankle gussets
  • Treated for limited flammability, tested in accordance with ASTM D-6413
  • Cape ventilated back with D-ring access slot for comfort and safety harness compatibility
Catalog No. 50-007-88423 Supplier MSC Supplier No. 51377588
This two-piece rain suit is engineered for applications requiring high visibility and protection against the elements. It features a jacket with an attached hood and elastic waist pants, designed to meet rigorous safety standards for high-visibility apparel.
  • Constructed from .40mm PU / Cotton Poly Blend stretch fabric
  • Meets ANSI/ISEA 107 2020 Type R Class 3 standards for high visibility
  • Features 2-inch silver reflective stripes
  • Jacket includes a zippered front with storm flap, two front patch pockets, and elastic inner cuffs
  • Pants feature an elastic waist with drawstring and expandable ankle gussets
  • Treated for limited flammability, tested in accordance with ASTM D-6413
  • Cape ventilated back with D-ring access slot for comfort and safety harness compatibility