Learn More
MSC MCR Safety Rain Suit, X-Large, Flame-Resistant, ANSI 107-2010, 1 Set
Catalog No. 5000768702
Encompass
This rain suit with bib overalls is designed for protection in wet and hazardous environments. It features flame-resistant properties and complies with multiple safety standards, making it suitable for demanding industrial applications.
  • Suit with bib overalls garment style
  • X-Large size
  • Black and lime colors
  • Polyester and polyurethane material
  • Disposable, flame-resistant, rain, and flame-retardant garment type
  • ANSI 107-2010, ASTM D6413-08, and NFPA 701 compliant
  • Attached hood
  • Open ankle style
  • Chest size of 56 in
  • Welt seam style
  • Elastic wrist style
  • Zipper closure
  • Plain front fly type
Catalog No. 50-007-68702 Supplier MSC Supplier No. 54579644
This rain suit with bib overalls is designed for protection in wet and hazardous environments. It features flame-resistant properties and complies with multiple safety standards, making it suitable for demanding industrial applications.
  • Suit with bib overalls garment style
  • X-Large size
  • Black and lime colors
  • Polyester and polyurethane material
  • Disposable, flame-resistant, rain, and flame-retardant garment type
  • ANSI 107-2010, ASTM D6413-08, and NFPA 701 compliant
  • Attached hood
  • Open ankle style
  • Chest size of 56 in
  • Welt seam style
  • Elastic wrist style
  • Zipper closure
  • Plain front fly type