Learn More
MCR Safety Rough Finish PVC Dipped Gloves
Catalog No. 19067303 Shop All MCR Safety Products
Change view
Click to view available options
Black; Size: 9.5
Catalog No.
19-067303 Black; Size: 9.5
1 options
This item is not returnable. View return policy
Catalog No. 19-067303 Supplier MCR Safety Supplier No. 6212R
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Single dipped Rough Finish PVC coated gloves provide basic protection in light chemical applications. Glove lengths from knit wrist up to 18 inches provide different levels of protection.
  • Breathable
  • Soft, seamless knit liner
  • Specifications
    Color Black
    Material PVC
    Size Large
    For Use With (Application) Agriculture, battery maintenance, chemical processing, fisheries, general maintenance, mild chemical protection, oil and gas industry
    Glove Cuff Style Gauntlet
    Liner Material Interlock Knit
    Lining Lined
    Length (English) 10.4 in.
    Length (Metric) 26.4 cm