Learn More
Description
- 15 mil, powder free latex
- Component materials comply with all federal regulations for food contact
- Industrial/Food Service Grade
- Textured Grip
- Double Chlorinated
Specifications
Specifications
| Certifications/Compliance | FDA |
| Applications | Industrial |
| Color | Green |
| Latex Free | Yes |
| Material | Latex |
| Product Type | Chemical and Liquid Protection Gloves |
| Size | 2X-Large |
| Thickness (English) | 15 mil |
| Thickness (Metric) | 0.38 mm |
| For Use With (Application) | Agriculture, battery maintenance, bottling, canning, dairy, food packing, mild chemical protection, paint manufacturing, painting, plating |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.