Learn More
MSC MCR Safety Safety Glasses, Anti-Fog, Blue Mirror, Full-Framed, 1 ea
Catalog No. 5000676369
Encompass
These safety glasses are designed with anti-fog, polycarbonate blue mirror lenses and a full-framed design. Featuring a black frame with non-adjustable temples, they are part of the Dominator DM2 Series and meet ANSI Z87.1+ standards. The frame and lens are made from polycarbonate, providing UV protection.
  • Anti-fog coatings improve visibility under hot and humid conditions
Catalog No. 50-006-76369 Supplier MSC Supplier No. 94632486
These safety glasses are designed with anti-fog, polycarbonate blue mirror lenses and a full-framed design. Featuring a black frame with non-adjustable temples, they are part of the Dominator DM2 Series and meet ANSI Z87.1+ standards. The frame and lens are made from polycarbonate, providing UV protection.
  • Anti-fog coatings improve visibility under hot and humid conditions