Learn More
MSC MCR Safety Safety Glasses, Anti-Fog, Gray Polycarbonate, Full-Framed, 1 ea
Catalog No. 5000674147
Encompass
These safety glasses feature an anti-fog coating for improved visibility in hot and humid conditions. They are full-framed with a wrap-around design and gray polycarbonate lenses, offering UV protection and meeting ANSI Z87.1+ standards.
  • Anti-fog coating for improved visibility
  • Full-framed, wrap-around design
  • Gray polycarbonate lenses
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Thermoplastic rubber frame material
Catalog No. 50-006-74147 Supplier MSC Supplier No. 66600206
These safety glasses feature an anti-fog coating for improved visibility in hot and humid conditions. They are full-framed with a wrap-around design and gray polycarbonate lenses, offering UV protection and meeting ANSI Z87.1+ standards.
  • Anti-fog coating for improved visibility
  • Full-framed, wrap-around design
  • Gray polycarbonate lenses
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Thermoplastic rubber frame material