Learn More
Enterprise Technology Solutions MCR Safety Safety Glasses, Anti-Fog, Over Glass, Clear
SDP
Catalog No. 5000828428
Encompass_Preferred
Anti-fog safety glasses feature an over-the-glass, wraparound lens design to protect your prescription eyewear and your eyes. Thermal plastic rubber temple inserts enhance the grip and comfort of these glasses. Built-in side protection prevents particles from entering from the sides.
Catalog No. 50-008-28428 Supplier Enterprise Technology Solutions Supplier No. MCSCRWOG110AF
Anti-fog safety glasses feature an over-the-glass, wraparound lens design to protect your prescription eyewear and your eyes. Thermal plastic rubber temple inserts enhance the grip and comfort of these glasses. Built-in side protection prevents particles from entering from the sides.