Learn More
MSC MCR Safety Safety Glasses, Anti-Fog, Polycarbonate, ANSI Z87.1, 1 ea
Catalog No. 5000793604
Encompass
These safety glasses feature anti-fog, polycarbonate, clear, and frameless lenses, designed for various applications requiring eye protection.
  • Anti-fog lens coating improves visibility in hot and humid conditions
  • Polycarbonate lens material for durability
  • Clear lenses and frameless design
  • Wrap-around frame provides enhanced coverage
  • Universal size for broad applicability
  • Meets ANSI Z87.1+ standard for eye protection
  • UV protection
  • Bayonet temple type
Catalog No. 50-007-93604 Supplier MSC Supplier No. 90174640
These safety glasses feature anti-fog, polycarbonate, clear, and frameless lenses, designed for various applications requiring eye protection.
  • Anti-fog lens coating improves visibility in hot and humid conditions
  • Polycarbonate lens material for durability
  • Clear lenses and frameless design
  • Wrap-around frame provides enhanced coverage
  • Universal size for broad applicability
  • Meets ANSI Z87.1+ standard for eye protection
  • UV protection
  • Bayonet temple type