Learn More
MSC MCR Safety Safety Glasses, Clear, Anti-Fog, Z87 Compliant, 1 ea
Catalog No. 5000788827
Encompass
MCR Safety SS1 series safety glasses offer a cost-efficient, single-lens design with a stylish wrap-around form factor, providing a wide area of protection and unrestricted vision. These glasses feature rugged polycarbonate lenses with UV-AF Anti-Fog coating.
  • Cost-efficient design
  • Wrap-around lens design
  • Built-in side shields
  • Extendable length spatula temples
  • One-piece polycarbonate lens
  • Integral side shields filter out 99.9% of UV radiation
  • Molded-in nose bridge
  • Lightweight frame
  • Clear lens with UV-AF Anti-Fog coating
  • ANSI/ISEA Z87.1-2025 and CE EN166 compliant
  • TAA compliant
Catalog No. 50-007-88827 Supplier MSC Supplier No. 52407905
MCR Safety SS1 series safety glasses offer a cost-efficient, single-lens design with a stylish wrap-around form factor, providing a wide area of protection and unrestricted vision. These glasses feature rugged polycarbonate lenses with UV-AF Anti-Fog coating.
  • Cost-efficient design
  • Wrap-around lens design
  • Built-in side shields
  • Extendable length spatula temples
  • One-piece polycarbonate lens
  • Integral side shields filter out 99.9% of UV radiation
  • Molded-in nose bridge
  • Lightweight frame
  • Clear lens with UV-AF Anti-Fog coating
  • ANSI/ISEA Z87.1-2025 and CE EN166 compliant
  • TAA compliant