Learn More
MSC MCR Safety Safety Glasses, Clear, Scratch-Resistant, Polycarbonate, Frameless, 1 ea
Catalog No. 5000787075
Encompass
MCR Safety Checklite CL4 series safety glasses are designed to provide solution-based eyewear protection with premium features at an affordable price. They feature a stylish hollowed-out lightweight frame for maximum airflow, suitable for everyday use or as visitor eyewear.
  • Scratch-resistant clear polycarbonate lenses
  • Frameless, wrap-around design
  • Non-adjustable clear bayonet temples with earplug retaining technology
  • UV protection
  • Meets ANSI Z87.1+ standards
Catalog No. 50-007-87075 Supplier MSC Supplier No. 95237756
MCR Safety Checklite CL4 series safety glasses are designed to provide solution-based eyewear protection with premium features at an affordable price. They feature a stylish hollowed-out lightweight frame for maximum airflow, suitable for everyday use or as visitor eyewear.
  • Scratch-resistant clear polycarbonate lenses
  • Frameless, wrap-around design
  • Non-adjustable clear bayonet temples with earplug retaining technology
  • UV protection
  • Meets ANSI Z87.1+ standards