Learn More
MSC MCR Safety Safety Glasses, Green Polycarbonate, ANSI Z87.1, 1 ea.
Catalog No. 5000787380
Encompass
These full-framed safety glasses are designed for welding applications, featuring scratch-resistant, polycarbonate green lenses and a green frame. They provide reliable eye protection and meet ANSI Z87.1+ standards.
  • Scratch-resistant lenses
  • Full-framed design
  • Wrap around frame
  • Green polycarbonate lenses
  • Green frame and temples
  • Non-adjustable temples
  • UV protection
  • Meets ANSI Z87.1+ standards
Catalog No. 50-007-87380 Supplier MSC Supplier No. 52408325
These full-framed safety glasses are designed for welding applications, featuring scratch-resistant, polycarbonate green lenses and a green frame. They provide reliable eye protection and meet ANSI Z87.1+ standards.
  • Scratch-resistant lenses
  • Full-framed design
  • Wrap around frame
  • Green polycarbonate lenses
  • Green frame and temples
  • Non-adjustable temples
  • UV protection
  • Meets ANSI Z87.1+ standards