Learn More
MSC MCR Safety Safety Glasses, MAX6 Anti-Fog, 1.5 Magnifier, 1 ea
Catalog No. 5000786944
Encompass
MCR Safety Checklite CL5 Safety Glasses provide solution-based eye protection with premium features at an affordable price. These glasses include a soft, closed-cell foam lining for enhanced eye protection and a lightweight frame for maximum airflow. They are designed for everyday use or as visitor specifications and meet ANSI/ISEA Z87.1 standards.
  • Earplug retaining technology
  • Foam lining for added protection
  • Hollowed-out, lightweight frame for maximum airflow
  • Removable closed-cell foam gasket
  • Bayonet temple for greater comfort
  • MAX6 anti-fog lens coating
  • Magnification of 1.5
  • U6 rated, CSA Z94.3, TAA compliant
Catalog No. 50-007-86944 Supplier MSC Supplier No. 98706682
MCR Safety Checklite CL5 Safety Glasses provide solution-based eye protection with premium features at an affordable price. These glasses include a soft, closed-cell foam lining for enhanced eye protection and a lightweight frame for maximum airflow. They are designed for everyday use or as visitor specifications and meet ANSI/ISEA Z87.1 standards.
  • Earplug retaining technology
  • Foam lining for added protection
  • Hollowed-out, lightweight frame for maximum airflow
  • Removable closed-cell foam gasket
  • Bayonet temple for greater comfort
  • MAX6 anti-fog lens coating
  • Magnification of 1.5
  • U6 rated, CSA Z94.3, TAA compliant