Learn More
MSC MCR Safety Safety Glasses, Uncoated, Polycarbonate, Clear, 1 ea
Catalog No. 5000782641
Encompass
These MCR Safety glasses are designed for over-the-glasses (OTG) use, providing clear vision and protection for various industrial applications. They feature a full-framed design with uncoated polycarbonate lenses.
  • Over-the-glasses (OTG) style
  • Uncoated clear polycarbonate lenses
  • Full-framed design with clear frame
  • Universal size with non-adjustable spatula temples
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Features side shield and wraparound style
Catalog No. 50-007-82641 Supplier MSC Supplier No. 61860664
These MCR Safety glasses are designed for over-the-glasses (OTG) use, providing clear vision and protection for various industrial applications. They feature a full-framed design with uncoated polycarbonate lenses.
  • Over-the-glasses (OTG) style
  • Uncoated clear polycarbonate lenses
  • Full-framed design with clear frame
  • Universal size with non-adjustable spatula temples
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Features side shield and wraparound style